Learn More
Keystone Cleanroom Products 1422A DuPont™ Tyvek™ Autoclave Sterilization Wrapper
Autoclavable Wrapper provides an excellent bacterial and microbial barrier for superior performance post steam sterilization.
£245.00 - £2725.00
Specifications
Product Type | Sterilization Wrap |
---|---|
Autoclavable | Autoclavable |
Certifications/Compliance | COC |
Color | White |
Disposable | Single-use |
Product Code | Brand | Dimensions (L x W) | No. per Case | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | No. per Case | Price | Quantity & Availability | ||||
17748517
|
Keystone Cleanroom Products
T2ASHEET01.5X01.5 |
1.5 x 1.5 in. | 500 |
£245.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17758517
|
Keystone Cleanroom Products
T2ASHEET0202 |
2 x 2 in. | 500 |
£245.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17768517
|
Keystone Cleanroom Products
T2ASHEET0303 |
3 x 3 in. | 500 |
£288.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17788517
|
Keystone Cleanroom Products
T2ASHEET0306 |
3 x 6 in. | 500 |
£338.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17798517
|
Keystone Cleanroom Products
T2ASHEET0404 |
4 x 4 in. | 500 |
£338.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17708527
|
Keystone Cleanroom Products
T2ASHEET0505 |
5 x 5 in. | 2000 |
£1845.00
Pack of 2000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17718527
|
Keystone Cleanroom Products
T2ASHEET0606 |
6 x 6 in. | 1000 |
£865.00
Pack of 1000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17728527
|
Keystone Cleanroom Products
T2ASHEET0608 |
6 x 8 in. | 1000 |
£1050.00
Pack of 1000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17738527
|
Keystone Cleanroom Products
T2ASHEET0609 |
6 x 9 in. | 1000 |
£1050.00
Pack of 1000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17748527
|
Keystone Cleanroom Products
T2ASHEET0612 |
6 x 12 in. | 1000 |
£1155.00
Pack of 1000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17758527
|
Keystone Cleanroom Products
T2ASHEET0624 |
6 x 24 in. | 500 |
£1155.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17768527
|
Keystone Cleanroom Products
T2ASHEET075075 |
7.5 x 7.5 in. | 1000 |
£865.00
Pack of 1000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17788527
|
Keystone Cleanroom Products
T2ASHEET0909 |
9 x 9 in. | 1000 |
£1040.00
Pack of 1000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17798527
|
Keystone Cleanroom Products
T2ASHEET0911 |
9 x 11 in. | 500 |
£550.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17708537
|
Keystone Cleanroom Products
T2ASHEET1010 |
10 x 10 in. | 500 |
£550.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17718537
|
Keystone Cleanroom Products
T2ASHEET1212 |
12 x 12 in. | 1000 |
£1270.00
Pack of 1000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17738537
|
Keystone Cleanroom Products
T2ASHEET1530 |
15 x 30 in. | 500 |
£1640.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17758537
|
Keystone Cleanroom Products
T2ASHEET1822 |
18 x 22 in. | 500 |
£1270.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17768537
|
Keystone Cleanroom Products
T2ASHEET2020 |
20 x 20 in. | 500 |
£1240.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17778537
|
Keystone Cleanroom Products
T2ASHEET2424 |
24 x 24 in. | 500 |
£1270.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17788537
|
Keystone Cleanroom Products
T2ASHEET3030 |
30 x 30 in. | 500 |
£1585.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17798537
|
Keystone Cleanroom Products
T2ASHEET3636 |
36 x 36 in. | 500 |
£2060.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17718547
|
Keystone Cleanroom Products
T2ASHEET4080 |
40 x 80 in. | 50 |
£421.00
Pack of 50 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17728547
|
Keystone Cleanroom Products
T2ASHEET4545 |
45 x 45 in. | 125 |
£1155.00
Pack of 125 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17738547
|
Keystone Cleanroom Products
T2ASHEET4848 |
48 x 48 in. | 125 |
£1155.00
Pack of 125 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17748547
|
Keystone Cleanroom Products
T2ASHEET5454 |
54 x 54 in. | 125 |
£1160.00
Pack of 125 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17758547
|
Keystone Cleanroom Products
T2ASHEET5472 |
54 x 72 in. | 125 |
£1580.00
Pack of 125 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17778527
|
Keystone Cleanroom Products
T2ASHEET0808 |
8 x 8 in. | 500 |
£475.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17748537
|
Keystone Cleanroom Products
T2ASHEET1818 |
18 x 18 in. | 500 |
£1000.00
Pack of 500 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
17778547
|
Keystone Cleanroom Products
T2ASHEET8494 |
84 x 94 in. | 125 |
£2725.00
Pack of 125 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||||
Description
Tyvek™ Autoclavable Wrappers are breathable while providing an excellent bacterial and microbial barrier for superior performance post steam sterilization.
- Lint-free (Helmke Drum) means no airborne particulates are generated during its use as sterilization packaging
- Clean and consistently cut using an automated CAD system per ISO 8 Requirements
- Packaged in an ISO 6 Controlled Clean Room
- Designed for Steam Sterilization
- Hydrophobic
- Resists tearing and punctures
Specifications
Sterilization Wrap | |
COC | |
Single-use | |
Pharmaceutical Production Equipment | |
Non-sterile | |
Steam, EtO, Gamma, VHP | |
<125°C |
Autoclavable | |
White | |
Steam Sterilization | |
HDPE | |
5.3 mm | |
<250°F |