Learn More
Axygen™ 10 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 384 Tip Format
For use with Agilent™/Velocity11 automation systems
Brand:  Axygen™ VT-384-10UL-R-S
Code : A0
Additional Details : Weight : 10.90000kg
 View more versions of this product
Product Code. 12551268
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
  • Clear polypropylene
  • Certified RNase-/DNase-free and nonpyrogenic
  • Nonpyrogenic
  • Maxymum Recovery™ technology for ultra-low retention
  • Available in nonsterile or sterile
Order Info
10 Racks/Pack, 5 Packs/Case
Specifications
Specifications
10 μL
Sterile
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
Automation, Round
50 Racks/Cs.
Polypropylene
Non-Filtered
Clear
DNase, Rnase Free
Rack
SBS Rack
Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST