missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ 10 μL Automation Tips for Agilent™/Velocity 11 VPrep™ and Bravo, 384 Tip Format
Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 19200
Pack of 3840
Description
Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
- Clear polypropylene
- Certified RNase-/DNase-free and nonpyrogenic
- Nonpyrogenic
- Maxymum Recovery™ technology for ultra-low retention
- Available in nonsterile or sterile
Order Info
10 Racks/Pack, 5 Packs/Case
Specifications
Specifications
| Volume (Metric) | 10 μL |
| Sterility | Non-sterile |
| Color | Clear |
| Clean Claims | DNase, RNase Free |
| For Use With (Application) | Non-pyrogenic, used for stringent specifications under strict process controls |
| Features | w/Double label |
| Format | Rack |
| Tip Style | Automation, Round |
| For Use With (Equipment) | Agilent/Velocity VPrep 16ST, Agilent/Velocity VPrep 96ST, Agilent/Velocity VPrep 384ST, Agilent Bravo 384ST |
| Material | Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction