accessibility menu, dialog, popup

UserName

H3COHNIHNCH3O

CAS RN 93515-00-5

IUPAC Name: N-[2-(2-iodo-5-methoxy-1H-indol-3-yl)ethyl]acetamide
Synonyms: N-[2-(2-iodo-5-methoxy-1H-indol-3-yl)ethyl]acetamide
Molecular Weight (g/mol): 358.18
Molecular Formula: C13H15IN2O2
InChi Key: FJDDSMSDZHURBJ-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC(I)=C(CCNC(C)=O)C2=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results