missing translation for 'onlineSavingsMsg'
Learn More

2-Iodomelatonin, Tocris Bioscience™
GREENER_CHOICE

Product Code. 30001397
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001397 10 mg 10mg
18085282 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001397 Supplier Tocris Bioscience Supplier No. 0737/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High affinity melatonin agonist

Potent melatonin agonist (pKi values are 10.55 and 9.87 at human recombinant MT1 and MT2 receptors respectively). 2-[125I]-iodomelatonin has been used to identify, characterize and localize melatonin binding sites in the brain and peripheral tissues.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93515-00-5
Molecular Formula C13H15IN2O2
Molecular Weight (g/mol) 358.179
InChI Key FJDDSMSDZHURBJ-UHFFFAOYSA-N
Synonym 2-iodomelatonin, n-2-2-iodo-5-methoxy-1h-indol-3-yl ethyl acetamide, 2-iodo-melatonin, 2-i-mlt, acetamide, n-2-2-iodo-5-methoxy-1h-indol-3-yl ethyl-, 3beta, acetamide,n-2-2-iodo-5-methoxy-1h-indol-3-yl ethyl, n-2-2-iodo-5-methoxyindol-3-yl ethyl acetamide, acetamide, n-2-2-iodo-5-methoxy-1h-indol-3-yl ethyl, melatonin,2-iodo
PubChem CID 115348
IUPAC Name N-[2-(2-iodo-5-methoxy-1H-indol-3-yl)ethyl]acetamide
SMILES CC(=O)NCCC1=C(NC2=C1C=C(C=C2)OC)I

Specifications

Quantity 10 mg
Formula Weight 358.18
Percent Purity >98%
Chemical Name or Material 2-Iodomelatonin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.