CAS RN 925681-41-0
IUPAC Name:
[4-(2-chloro-10H-phenoxazin-10-yl)butyl]diethylamine hydrochloride
Synonyms:
[4-(2-chlorophenoxazin-10-yl)butyl]diethylamine hydrochloride
Molecular Weight (g/mol):
381.34
Molecular Formula:
C20H26Cl2N2O
InChi Key:
SVKSJUIYYCQZEC-UHFFFAOYSA-N
SMILES:
Cl.CCN(CC)CCCCN1C2=CC=CC=C2OC2=C1C=C(Cl)C=C2
1
–
3
of
3
results
1
Tocris Bioscience™ 10-DEBC hydrochloride
CAS: 925681-41-0 Molecular Formula: C20H26Cl2N2O
2