missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Akt Inhibitor X

Product Code. 15426739
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15426739 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15426739 Supplier Thermo Scientific Chemicals Supplier No. J65119.#0
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

Akt Inhibitor X is used as a cell-permeable, reversible, and selective inhibitor of the phosphorylation of Akt and its in vitro kinase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A cell-permeable, reversible, and selective inhibitor of the phosphorylation of Akt and its in vitro kinase activity
CAS 925681-41-0
Molecular Formula C20H25ClN2O.HCl, C20H26Cl2N2O
Formula Weight 381.34
Synonym 2-Chloro-10-[4-(diethylamino)butyl]phenoxazine hydrochloride; 4-(2-Chloro-10H-phenoxazin-10-yl)-N,N-diethylbutan-1-amine hydrochloride
Sensitivity Light Sensitive and Hygroscopic
Solubility Soluble to 100mM in water or DMSO
Product Type Akt Inhibitor X
Form Powder
InChI Key SVKSJUIYYCQZEC-UHFFFAOYSA-N
SMILES Cl.CCN(CC)CCCCN1C2=CC=CC=C2OC2=C1C=C(Cl)C=C2
IUPAC Name [4-(2-chloro-10H-phenoxazin-10-yl)butyl]diethylamine hydrochloride
Molecular Weight (g/mol) 381.34
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.