missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies AKT Inhibitor X (Hydrochloride)

Product Code. 17130152
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17130152 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17130152 Supplier STEMCELL Technologies Supplier No. 72952

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

PI3K/AKT pathway inhibitor; Inhibits AKT

  • AKT Inhibitor X is a cell-permeable phenoxazine-derivative inhibitor of AKT kinase phosphorylation with an IC50 of ∼1 - 2 μM
  • AKT Inhibitor X blocks translocation of AKT after insulin-like growth factor 1 (IGF-1) treatment (Thimmaiah et al.) This product is supplied as the hydrochloride salt of the molecule
  • Inhibits growth and induces apoptosis of human rhabdomyosarcoma cell lines (Thimmaiah et al.)
  • Inhibits proliferation of breast cancer cell lines, alone or synergistically with chloroquine (Hu et al.)
  • Reduces replication of Myxoma virus in a variety of human tumor cell lines (Werden & McFadden)
  • Induces autophagy in neurons and is neuroprotective in a primary neuronal Huntington Disease cellular model (Tsvetkov et al.)

Specifications

CAS 925681-41-0
Synonym 10-DEBC hydrochloride
Quantity 10 mg
Molecular Formula C20H25ClN2O · HCl
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.