accessibility menu, dialog, popup

UserName

CAS RN 71-55-6

CH3ClClCl

CAS RN 71-55-6

IUPAC Name: 1,1,1-trichloroethane
Synonyms: trichloroethane baltana tafclean caswell no. 875 ethana NU chlorothene dowclene LS chlorotene cleanite genklene LB
Molecular Weight (g/mol): 133.4
Molecular Formula: C2H3Cl3
InChi Key: UOCLXMDMGBRAIB-UHFFFAOYSA-N
SMILES: CC(Cl)(Cl)Cl
Solvent or Matrix 
  • (2)
  • (3)
  • (1)
Concentration 
  • (1)
  • (1)
Element 
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)