missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VOC-Mix 21 200 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| CAS | 71-55-6, 106-93-4, 107-06-2, 75-27-4, 67-66-3, 124-48-1 |
| Quantity | 1 mL |
| Concentration | Tetrachloromethane 199.6 μg/mL∣1,2-Dichloroethane 199.9 μg/mL∣1,1,1-Trichloroethane 200 μg/mL∣Chloroform 200 μg/mL∣Tetrachloroethene 200.1 μg/mL∣Tribromomethane 200.1 μg/mL∣Bromodichloromethane 200.2 μg/mL∣Dibromochloromethane 200.2 μg/mL∣1,2-Dibromoethane 200.4 μg/mL∣Trichloroethene 200.9 μg/mL |
| Certifications/Compliance | ISO/IEC 17025 |
| Solvent or Matrix | Methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction