missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VOC-Mix 7 2000 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| CAS | 71-55-6, 75-35-4, 107-06-2, 106-46-7, 71-43-2, 75-27-4 |
| Quantity | 1 mL |
| Concentration | 1,1,1-Trichloroethane 2000 μg/mL∣1,1-Dichloroethene 2000 μg/mL∣1,2-Dichloroethane 2000 μg/mL∣1,4-Dichlorobenzene 2000 μg/mL∣Benzene 2000 μg/mL∣Bromodichloromethane 2000 μg/mL∣Chloroform 2000 μg/mL∣Dibromochloromethane 2000 μg/mL∣Tetrachloromethane 2000 μg/mL∣Tribromomethane 2000 μg/mL∣Trichloroethene 2000 μg/mL |
| Certifications/Compliance | ISO/IEC 17025 |
| Solvent or Matrix | Methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction