missing translation for 'onlineSavingsMsg'
Learn More

VOC-Mix 9 1000 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30314284
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30314284

Brand: Dr. Ehrenstorfer DREYA05000009AC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

CAS 71-55-6, 71-43-2, 67-66-3, 156-59-2, 75-09-2, 108-38-3
Quantity 1 mL
Concentration 1,1,1-Trichloroethane 1000 μg/mL∣Benzene 1000 μg/mL∣Chloroform 1000 μg/mL∣cis-1,2-Dichloroethene 1000 μg/mL∣Dichloromethane (Methylenechloride) 1000 μg/mL∣m-Xylene (1,3-Dimethylbenzene) 1000 μg/mL∣o-Xylene (1,2-Dimethylbenzene) 1000 μg/mL∣p-Xylene (1,4-Dimethylbenzene) 1000 μg/mL∣Tetrachloroethene 1000 μg/mL∣Tetrachloromethane 1000 μg/mL∣Toluene 1000 μg/mL∣Trichloroethene 1000 μg/mL
Certifications/Compliance ISO/IEC 17025
Solvent or Matrix Acetone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.