accessibility menu, dialog, popup

UserName

CAS RN 64022-27-1

HClClNNNHN

CAS RN 64022-27-1

IUPAC Name: hydrogen 2-chloro-6-(piperazin-1-yl)pyrazine chloride
Synonyms: hydrogen 2-chloro-6-(piperazin-1-yl)pyrazine chloride
Molecular Weight (g/mol): 235.11
Molecular Formula: C8H12Cl2N4
InChi Key: PFIZGLUIYAZQFU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].ClC1=CN=CC(=N1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results