missing translation for 'onlineSavingsMsg'
Learn More

MK 212 hydrochloride, Tocris Bioscience™
GREENER_CHOICE

Product Code. 30001152
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001152 10 mg 10mg
18074592 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001152 Supplier Tocris Bioscience Supplier No. 0941/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

5-HT2C agonist

5-HT receptor agonist; displays selectivity for 5-HT2C over 5-HT2A (IC50 values are 0.028 and 0.42 μM for human 5-HT2C and 5-HT2A receptors expressed in HEK293 cells respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64022-27-1
Molecular Formula C8H12Cl2N4
Molecular Weight (g/mol) 235.11
MDL Number MFCD04975444
InChI Key PFIZGLUIYAZQFU-UHFFFAOYSA-N
Synonym 2-chloro-6-1-piperazinyl pyrazine, 6-chloro-2-1-piperazinyl pyrazine, 2-chloro-6-piperazin-1-yl pyrazine, pyrazine, 2-chloro-6-1-piperazinyl, tocris-0941, biomol-nt_000130, d02esw, gtpl165, 1-6-chloro-2-pyrazinyl piperazine, 6-chloro-2-1-piperazinyl-pyrazine
PubChem CID 107992
IUPAC Name hydrogen 2-chloro-6-(piperazin-1-yl)pyrazine chloride
SMILES [H+].[Cl-].ClC1=CN=CC(=N1)N1CCNCC1

Specifications

Quantity 10 mg
Formula Weight 235.12
Percent Purity >99%
Chemical Name or Material MK 212 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.