accessibility menu, dialog, popup

UserName

CAS RN 6323-79-1

CH3NH3CCH

CAS RN 6323-79-1

IUPAC Name: (prop-2-yn-1-yl)dipropylamine
Synonyms: prop-2-yn-1-yldipropylamine
Molecular Weight (g/mol): 139.24
Molecular Formula: C9H17N
InChi Key: ATTMKPWGJILFFV-UHFFFAOYSA-N
SMILES: CCCN(CCC)CC#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results