missing translation for 'onlineSavingsMsg'
Learn More

3-Di-n-propylamino-1-propyne, 98%, Thermo Scientific™

Product Code. 11362537
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11362537

Brand: Thermo Scientific Alfa Aesar L11633.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6323-79-1
Molecular Formula C9H17N
Molecular Weight (g/mol) 139.242
MDL Number MFCD00041626
InChI Key ATTMKPWGJILFFV-UHFFFAOYSA-N
Synonym 3-di-n-propylamino-1-propyne, n,n-dipropyl-2-propyn-1-amine, propargyldipropylamine, n,n-dipropylprop-2-yn-1-amine, 3-dipropylamino-1-propyne, 2-propyn-1-amine, n,n-dipropyl, prop-2-ynyldipropylamine, dipropylpropargylamine, propargyl di-n-propyl amine, n,n-dipropyl-2-propynylamine
PubChem CID 138724
IUPAC Name N-propyl-N-prop-2-ynylpropan-1-amine
SMILES CCCN(CCC)CC#C

Specifications

Density 0.828
Boiling Point 157°C to 159°C
Flash Point 55°C (131°F)
Odor Amine-like
Refractive Index 1.435
Quantity 5 g
UN Number UN2733
Beilstein 1746300
Formula Weight 139.24
Percent Purity 98%
Chemical Name or Material 3-Di-n-propylamino-1-propyne
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.