missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lidocaine Hydrochloride Monohydrate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Lidocaine Hydrochloride Monohydrate
Specifications
Specifications
| Chemical Name or Material | Lidocaine hydrochloride monohydrate |
| CAS | 6108-05-0 |
| Analyte or Component Names | Lidocaine hydrochloride monohydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H22 N2 O . Cl H . H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Lidocaine Hydrochloride Monohydrate, CCI 1873A |
| InChI Formula | InChI=1S/C14H22N2O.ClH.H2O/c1-5-16(6-2)10-13(17)15-14-11(3)8-7-9-12(14)4;;/h7-9H,5-6,10H2,1-4H3,(H,15,17);1H;1H2 |
| SMILES | O.Cl.CCN(CC)CC(=O)Nc1c(C)cccc1C |
| IUPAC Name | 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide;hydrate;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction