missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine Hydrochloride Monohydrate, TRC

Product Code. 30135825
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135825 10 g 10g
30132442 100 g 100g
30040789 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135825 Supplier Toronto Research Chemicals Supplier No. TRCL39780010G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6108-05-0
Molecular Formula C14 H22 N2 O . Cl H . H2 O
Molecular Weight (g/mol) 288.81
Synonym Lidocaine hydrochloride monohydrate, Acetamide, 2-(diethylamino)-N-(2,6-dimethylphenyl)-, hydrochloride, hydrate (1:1:1), 2',6'-Acetoxylidide, 2-(diethylamino)-, monohydrochloride, monohydrate (8CI), Acetamide, 2-(diethylamino)-N-(2,6-dimethylphenyl)-, monohydrochloride, monohydrate (9CI), Lidocaine monohydrate hydrochloride, Lidocaine monohydrochloride monohydrate, Petercaine, α-Diethylamino-2,6-acetoxylidide hydrochloride monohydrate, 2-(Diethylamino)-N-(2,6-dimethylphenyl)acetamide hydrochloride monohydrate, 2-(Diethylamino)-2',6'-acetoxylidide hydrochloride monohydrate
IUPAC Name 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide;hydrate;hydrochloride
SMILES O.Cl.CCN(CC)CC(=O)Nc1c(C)cccc1C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H22N2O.ClH.H2O/c1-5-16(6-2)10-13(17)15-14-11(3)8-7-9-12(14)4;;/h7-9H,5-6,10H2,1-4H3,(H,15,17);1H;1H2
Formula Weight 288.1605
Chemical Name or Material Lidocaine hydrochloride monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.