accessibility menu, dialog, popup

UserName

CAS RN 582323-16-8

ONHNNClFF

CAS RN 582323-16-8

IUPAC Name: N-(2-chloropyrimidin-5-yl)-3,4-difluorobenzamide
Synonyms: N-(2-chloropyrimidin-5-yl)-3,4-difluorobenzamide
Molecular Weight (g/mol): 269.64
Molecular Formula: C11H6ClF2N3O
InChi Key: IIBSHMFXVWTQSJ-UHFFFAOYSA-N
SMILES: FC1=C(F)C=C(C=C1)C(=O)NC1=CN=C(Cl)N=C1
Grade 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results