missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ICA 069673

Description
ICA 069673 is a selective KV7.2/KV7.3 (KCNQ2/3) channel opener (EC50 = 0.69 μM). Exhibits 20-fold selectivity for KV7.2/KV7.3 over KV7.3/KV7.5, with no measurable activity over a panel of cardiac ion channels. Hyperpolarizes resting membrane potential and inhibits spontaneous action potentials in guinea pig detrusor muscle (DSM) isolated cells. Orally active in animal models of epilepsy.
Specifications
Specifications
| Chemical Name or Material | N-(2-Chloro-5-pyrimidinyl)-3,4-difluorobenzamide |
| CAS | 582323-16-8 |
| Assay Percent Range | >99% |
| Quantity | 5 mg |
| Molecular Formula | C11H6ClF2N3O |
| Formula Weight | Observed MW: 269.63 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?