missing translation for 'onlineSavingsMsg'
Learn More

ICA-069673, MedChemExpress

Product Code. 16496067
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496067 5 mg 5mg
16484586 100 mg 100mg
16479876 50 mg 50mg
16472726 25 mg 25mg
16423348 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496067 Supplier Medchem Express Supplier No. HY101396

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ICA-069673 is a KCNQ2/Q3 potassium channel activator with an IC50 of 0.69 μM.

ICA-069673 is a KCNQ2/Q3 potassium channel activator with an IC50 of 0.69 μM.

Specifications

Chemical Name or Material ICA-069673
CAS 582323-16-8
Color White
Purity Grade Notes Research
Molecular Formula C11H6ClF2N3O
Quantity 5 mg
Solubility Information DMSO : ≥ 34 mg/mL (126.10 mM)
SMILES O=C(NC1=CN=C(Cl)N=C1)C2=CC=C(F)C(F)=C2
Molecular Weight (g/mol) 269.63
Formula Weight 269.63
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.