accessibility menu, dialog, popup

UserName

NH2(S)NOHO

CAS RN 57213-48-6

IUPAC Name: (2S)-2-azaniumyl-3-(3-cyanophenyl)propanoate
Synonyms: (2S)-2-ammonio-3-(3-cyanophenyl)propanoate
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: ZHUOMTMPTNZOJE-VIFPVBQESA-N
SMILES: [NH3+][C@@H](CC1=CC=CC(=C1)C#N)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)