missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-L-phenylalanine, 98%

Product Code. 15413697
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15413697 250 mg 250mg
11307359 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15413697 Supplier Thermo Scientific Alfa Aesar Supplier No. H27474.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 57213-48-6 | C10H10N2O2 | 190.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57213-48-6
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.20
MDL Number MFCD01860885
InChI Key ZHUOMTMPTNZOJE-VIFPVBQESA-N
Synonym l-3-cyanophenylalanine, 3-cyano-l-phenylalanine, h-phe 3-cn-oh, s-2-amino-3-3-cyanophenyl propanoic acid, 2s-2-amino-3-3-cyanophenyl propanoic acid, phenylalanine, 3-cyano, l-3-cyano-phenylalanine, 3-cyanophenylalanine, pubchem15793
PubChem CID 2734509
IUPAC Name (2S)-2-azaniumyl-3-(3-cyanophenyl)propanoate
SMILES N[C@@H](CC1=CC=CC(=C1)C#N)C(O)=O

Specifications

UN Number UN3439
Quantity 250 mg
Formula Weight 190.2
Percent Purity 98%
Chemical Name or Material 3-Cyano-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.