missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (S)-N-3-Cyanophenylalanine, 95%, 98% ee

Product Code. 10592802 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10592802 5 g Glass bottle 5g
10394592 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10592802 Supplier Thermo Scientific Chemicals Supplier No. 362060050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 57213-48-6 | C10H10N2O2 | 190.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57213-48-6
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.20
MDL Number MFCD01860885
InChI Key ZHUOMTMPTNZOJE-VIFPVBQESA-N
Synonym l-3-cyanophenylalanine, 3-cyano-l-phenylalanine, h-phe 3-cn-oh, s-2-amino-3-3-cyanophenyl propanoic acid, 2s-2-amino-3-3-cyanophenyl propanoic acid, phenylalanine, 3-cyano, l-3-cyano-phenylalanine, 3-cyanophenylalanine, pubchem15793
PubChem CID 2734509
IUPAC Name (2S)-2-amino-3-(3-cyanophenyl)propanoic acid
SMILES N[C@@H](CC1=CC=CC(=C1)C#N)C(O)=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White to Yellow
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 190.2
Percent Purity 95%,98% ee
Physical Form Powder
Chemical Name or Material (S)-N-3-Cyanophenylalanine, 95%, (98% e.e.)
Show More Show Less
compliance-icons
Product Identifier
  • (S)-N-3-Cyanophenylalanine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.