accessibility menu, dialog, popup

UserName

CAS RN 56824-91-0

CH3OONClCl

CAS RN 56824-91-0

IUPAC Name: ethyl 4,8-dichloroquinoline-3-carboxylate
Synonyms: ethyl 4,8-dichloroquinoline-3-carboxylate
Molecular Weight (g/mol): 270.11
Molecular Formula: C12H9Cl2NO2
InChi Key: HINLPSGAVZPTIW-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C2C(Cl)=CC=CC2=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results