accessibility menu, dialog, popup

UserName

CAS RN 5465-65-6

H3COClNOO

CAS RN 5465-65-6

IUPAC Name: 1-(4-chloro-3-nitrophenyl)ethan-1-one
Synonyms: 1-(4-chloro-3-nitrophenyl)ethanone
Molecular Weight (g/mol): 199.59
Molecular Formula: C8H6ClNO3
InChi Key: YEVPHFIFGUWSMG-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results