missing translation for 'onlineSavingsMsg'
Learn More

4'-Chloro-3'-nitroacetophenone, 97%, Thermo Scientific™

Product Code. 11492078
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11492078

Brand: Thermo Scientific Alfa Aesar A15121.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5465-65-6
Molecular Formula C8H6ClNO3
Molecular Weight (g/mol) 199.59
MDL Number MFCD00007083
InChI Key YEVPHFIFGUWSMG-UHFFFAOYSA-N
Synonym 4'-chloro-3'-nitroacetophenone, 1-4-chloro-3-nitrophenyl ethanone, 4-chloro-3-nitroacetophenone, ethanone, 1-4-chloro-3-nitrophenyl, 1-4-chloro-3-nitrophenyl ethan-1-one, unii-gkv85f2zxl, acetophenone, 4'-chloro-3'-nitro, gkv85f2zxl, 4-chloro-3'-nitroacetophenone, 3'-nitro-4'-chloro acetophenone
PubChem CID 79596
IUPAC Name 1-(4-chloro-3-nitrophenyl)ethanone
SMILES CC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Melting Point 96°C to 100°C
Quantity 5 g
Beilstein 1640627
Formula Weight 199.6
Percent Purity 97%
Chemical Name or Material 4'-Chloro-3'-nitroacetophenone
compliance-icons
Product Identifier
  • 4'-Chloro-3'-nitroacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 4'-Chloro-3'-nitroacetophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.