accessibility menu, dialog, popup

UserName

CAS RN 532-03-6

CH3OOOHONH2O

CAS RN 532-03-6

IUPAC Name: 2-hydroxy-3-(2-methoxyphenoxy)propyl carbamate
Synonyms: methocarbamol (inn)
Molecular Weight (g/mol): 241.24
Molecular Formula: C11H15NO5
InChi Key: GNXFOGHNGIVQEH-UHFFFAOYNA-N
SMILES: COC1=CC=CC=C1OCC(O)COC(N)=O
Certifications/Compliance 
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results