missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methocarbamol, LoGiCal
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
missing translation for 'unitSize'
10mg
Specifications
Specifications
| Chemical Name or Material | Methocarbamol |
| CAS | 532-03-6 |
| Analyte or Component Names | Methocarbamol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H15 N O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 3-(o-Methoxyphenoxy)-1,2-propanediol-1-carbamate, Carbamic acid 2-Hydroxy-3-(o-methoxyphenoxy)propyl ester, 2-Hydroxy-3-(o-methoxyphenoxy)propyl carbamate, 3-(2-Methoxyphenoxy)-1-glyceryl carbamate, 3-(o-Methoxyphenoxy)-1,2-propanediol 1-carbamate, 3-(o-Methoxyphenoxy)-2-hydroxypropyl carbamate, AHR 85, Avetil, Delaxin, Etroflex, Glycerylguaiacolate carbamate, Guaiacol glyceryl ether carbamate, Guaiphenesin carbamate, Lumirelax, Methocarbamol, Miolaxene, NSC 170960, Neuraxin, Reflexyn, Relestrid, Robamol, Robaxin, Robinax, Surquetil, Traumacut, Tresortil, guaiamar carbamate, racemic-Methocarbamol |
| InChI Formula | InChI=1S/C11H15NO5/c1-15-9-4-2-3-5-10(9)16-6-8(13)7-17-11(12)14/h2-5,8,13H,6-7H2,1H3,(H2,12,14) |
| Solution Type | Neat |
| SMILES | COc1ccccc1OCC(O)COC(=O)N |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction