missing translation for 'onlineSavingsMsg'
Learn More

Methocarbamol, MedChemExpress

Product Code. 16478196
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16478196

Brand: Medchem Express HYB0262

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Methocarbamol is an orally active central muscle relaxant and blocks muscular Nav1.4 channel. Methocarbamol reversibly affects voltage dependence of inactivation of Nav1.4 channel. Methocarbamol has the potential for muscle spasms and pain syndromes research.

Methocarbamol is an orally active central muscle relaxant and blocks muscular Nav1.4 channel. Methocarbamol reversibly affects voltage dependence of inactivation of Nav1.4 channel. Methocarbamol has the potential for muscle spasms and pain syndromes research.

Specifications

Chemical Name or Material Methocarbamol
CAS 532-03-6
Color White
Purity Grade Notes Research
Molecular Formula C11H15NO5
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (414.52 mM) ∣H2O : 4 mg/mL (16.58 mM; Need ultrasonic)
SMILES OC(COC1=CC=CC=C1OC)COC(N)=O
Molecular Weight (g/mol) 241.24
Formula Weight 241.24
Percent Purity 98.39%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.