accessibility menu, dialog, popup

UserName

CAS RN 53100-44-0

CH3H3CCH3OON(S)OOHO

CAS RN 53100-44-0

IUPAC Name: (2S)-1-[(tert-butoxy)carbonyl]-5-oxopyrrolidine-2-carboxylic acid
Synonyms: (2S)-1-(tert-butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 229.23
Molecular Formula: C10H15NO5
InChi Key: MJLQPFJGZTYCMH-LURJTMIESA-N
SMILES: CC(C)(C)OC(=O)N1[C@@H](CCC1=O)C(O)=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)