missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boc-Pyr-OH, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Boc-Pyr-OH |
| CAS | 53100-44-0 |
| Molecular Formula | C10 H15 N O5 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C10H15NO5/c1-10(2,3)16-9(15)11-6(8(13)14)4-5-7(11)12/h6H,4-5H2,1-3H3,(H,13,14)/t6-/m0/s1 |
| SMILES | CC(C)(C)OC(=O)N1[C@@H](CCC1=O)C(=O)O |
| IUPAC Name | (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-5-oxopyrrolidine-2-carboxylic acid |
| Molecular Weight (g/mol) | 229.23 |
| Formula Weight | 229.095 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction