CAS RN 521984-48-5
CAS RN 521984-48-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
1,2,3,4-Tetrahydro-6,7-dimethoxy-2-[(2E)-3-(1-methyl-2-phenyl-1H-pyrrolo[2,3-b]pyridin-3-yl)-1-oxo-2-propenyl]-isoquinoline Monohydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1,2,3,4-Tetrahydro-6,7-dimethoxy-2-[(2E)-3-(1-methyl-2-phenyl-1H-pyrrolo[2,3-b]pyridin-3-yl)-1-oxo-2-propenyl]-isoquinoline Monohydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1,2,3,4-Tetrahydro-6,7-dimethoxy-2-[(2E)-3-(1-methyl-2-phenyl-1H-pyrrolo[2,3-b]pyridin-3-yl)-1-oxo-2-propenyl]-isoquinoline Monohydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
(E)-SIS3, MedChemExpress
MedChemExpress (E)-SIS3 is a potent and selective inhibitor of Smad3 with an IC50 of 3 μM for Smad3 phosphorylation. (E)-SIS3 inhibits the myofibroblast differentiation of fibroblasts by TGF-β1.