missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(E)-SIS3, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
Description
(E)-SIS3 is a potent and selective inhibitor of Smad3 with an IC50 of 3 μM for Smad3 phosphorylation. (E)-SIS3 inhibits the myofibroblast differentiation of fibroblasts by TGF-β1.
Specifications
Specifications
| Chemical Name or Material | (E)-SIS3 |
| CAS | 521984-48-5 |
| Color | Deep Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C28H28ClN3O3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 125 mg/mL (255.11 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | [H]Cl.O=C(/C=C/C1=C(N(C2=NC=CC=C21)C)C3=CC=CC=C3)N4CC5=C(CC4)C=C(C(OC)=C5)OC.[E] |
| Molecular Weight (g/mol) | 489.99 |
| Formula Weight | 489.99 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction