missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2,3,4-Tetrahydro-6,7-dimethoxy-2-[(2E)-3-(1-methyl-2-phenyl-1H-pyrrolo[2,3-b]pyridin-3-yl)-1-oxo-2-propenyl]-isoquinoline Monohydrochloride, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | 1,2,3,4-Tetrahydro-6,7-dimethoxy-2-[(2E)-3-(1-methyl-2-phenyl-1H-pyrrolo[2,3-b]pyridin-3-yl)-1-oxo-2-propenyl]-isoquinoline Monohydrochloride |
| CAS | 521984-48-5 |
| Molecular Formula | C28 H27 N3 O3 . H Cl |
| Quantity | 5 mg |
| Synonym | SIS 3 |
| InChI Formula | InChI=1S/C28H27N3O3.ClH/c1-30-27(19-8-5-4-6-9-19)22(23-10-7-14-29-28(23)30)11-12-26(32)31-15-13-20-16-24(33-2)25(34-3)17-21(20)18-31;/h4-12,14,16-17H,13,15,18H2,1-3H3;1H/b12-11+; |
| SMILES | Cl.COc1cc2CCN(Cc2cc1OC)C(=O)\C=C\c3c(c4ccccc4)n(C)c5ncccc35 |
| IUPAC Name | (E)-1-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)-3-(1-methyl-2-phenylpyrrolo[2,3-b]pyridin-3-yl)prop-2-en-1-one;hydrochloride |
| Molecular Weight (g/mol) | 489.993 |
| Formula Weight | 489.182 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction