accessibility menu, dialog, popup

UserName

CAS RN 488-10-8

CH3CH3O(Z)

CAS RN 488-10-8

IUPAC Name: 3-methyl-2-[(2Z)-pent-2-en-1-yl]cyclopent-2-en-1-one
Synonyms: jasmone cis-jasmone (Z)-jasmone
Molecular Weight (g/mol): 164.25
Molecular Formula: C11H16O
InChi Key: XMLSXPIVAXONDL-PLNGDYQASA-N
SMILES: CC\C=C/CC1=C(C)CCC1=O
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results