missing translation for 'onlineSavingsMsg'
Learn More

cis-Jasmone, 94%, Thermo Scientific™

Product Code. 11429366
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11429366

Brand: Thermo Scientific Alfa Aesar A19060.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-10-8
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00001402,MFCD00001402,MFCD00001402
InChI Key XMLSXPIVAXONDL-PLNGDYQASA-N
Synonym trans-jasmone, e-jasmone, unii-7tcs3y45dr, 7tcs3y45dr, 3-methyl-2-e-pent-2-enyl cyclopent-2-en-1-one, 2-cyclopenten-1-one, 3-methyl-2-2-pentenyl-, e, 3-methyl-2-pent-2-en-1-yl cyclopent-2-en-1-one, 3-methyl-2-2e-pent-2-en-1-yl cyclopent-2-en-1-one, jasmone, e, 3-methyl-2-2-pentenyl-2-cyclopenten-1-one
PubChem CID 1549019
IUPAC Name 3-methyl-2-[(2Z)-pent-2-en-1-yl]cyclopent-2-en-1-one
SMILES CC\C=C/CC1=C(C)CCC1=O

Specifications

Density 0.94
Boiling Point 134°C to 135°C (12 mmHg)
Flash Point 107°C (224°F)
Odor Pleasant
Refractive Index 1.499
Quantity 100 g
Beilstein 1907713
Merck Index 14,5259
Formula Weight 164.25
Percent Purity 94%
Chemical Name or Material cis-Jasmone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.