accessibility menu, dialog, popup

UserName

CAS RN 4500-12-3

HON(?)

CAS RN 4500-12-3

IUPAC Name: N-(adamantan-2-ylidene)hydroxylamine
Synonyms: 2-adamantanone, oxime
Molecular Weight (g/mol): 165.24
Molecular Formula: C10H15NO
InChi Key: RABVIFXMFZFITE-KHPPLWFESA-N
SMILES: ON=C1C2CC3CC(C2)CC1C3
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results