missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanone oxime, 97%, Thermo Scientific™

Product Code. 15434548
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 15434548

Brand: Thermo Scientific Alfa Aesar H58220.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4500-12-3
MDL Number MFCD00078273

Specifications

Molecular Formula C10H15NO
Quantity 10 g
Sensitivity Moisture sensitive
Synonym 2-adamantanone oxime, adamantan-2-one oxime, adamantanone oxime, 2-adamantanone, oxime, 2-hydroxyiminoadamantane, adamantanone-2-oxime, 2-isonitrosoadamantane, 2-hydroxyimino adamantane, n-2-adamantylidene hydroxylamine, n-adamantan-2-ylidene hydroxylamine
InChI Key RABVIFXMFZFITE-KHPPLWFESA-N
SMILES C1C2CC3CC1CC(C2)C3=NO
IUPAC Name N-(2-adamantylidene)hydroxylamine
Molecular Weight (g/mol) 165.24
PubChem CID 64158
Formula Weight 165.24
Percent Purity 97%
Chemical Name or Material 2-Adamantanone oxime
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.