CAS RN 393514-24-4
CAS RN 393514-24-4
IUPAC Name:
N-(4-tert-butylphenyl)-4-(3-chloropyridin-2-yl)piperazine-1-carboxamide
Synonyms:
N-(4-tert-butylphenyl)-4-(3-chloropyridin-2-yl)piperazine-1-carboxamide
Molecular Weight (g/mol):
372.9
Molecular Formula:
C20H25ClN4O
InChi Key:
ROGUAPYLUCHQGK-UHFFFAOYSA-N
SMILES:
CC(C)(C)C1=CC=C(NC(=O)N2CCN(CC2)C2=C(Cl)C=CC=N2)C=C1
1
–
2
of
2
results
1
Tocris Bioscience™ BCTC
CAS: 393514-24-4 Molecular Formula: C20H25ClN4O
2
BCTC, MedChemExpress
MedChemExpress BCTC is a potent and specific inhibitor of transient receptor potential cation channel subfamily M member 8 (TRPM8) in prostate cancer (PCa) DU145 cells.