missing translation for 'onlineSavingsMsg'
Learn More

BCTC, MedChemExpress

Product Code. p-200101880
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493907

Brand: Medchem Express HY19960

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BCTC is a potent and specific inhibitor of transient receptor potential cation channel subfamily M member 8 (TRPM8) in prostate cancer (PCa) DU145 cells.

BCTC is a potent and specific inhibitor of transient receptor potential cation channel subfamily M member 8 (TRPM8) in prostate cancer (PCa) DU145 cells.

Specifications

Chemical Name or Material BCTC
CAS 393514-24-4
Color White
Purity Grade Notes Research
Molecular Formula C20H25ClN4O
Quantity 50 mg
Solubility Information DMSO : ≥ 50 mg/mL (134.09 mM)
SMILES O=C(N1CCN(C2=NC=CC=C2Cl)CC1)NC3=CC=C(C(C)(C)C)C=C3
Molecular Weight (g/mol) 372.89
Formula Weight 372.89
Percent Purity 97.23%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.