missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BCTC
GREENER_CHOICE

Product Code. 30002115
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30002115

Brand: Tocris Bioscience™ 3875/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TRPV1 antagonist

Orally effective vanilloid receptor 1 (TRPV1 receptor) antagonist. Inhibits acid- and capsaicin-induced activation of rat TRPV1 receptors (IC50 values are 6.0 and 35 nM respectively). Displays analgesic properties in rat models of inflammatory and neuropathic pain. CNS penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393514-24-4
Molecular Formula C20H25ClN4O

Specifications

Quantity 10 mg
Formula Weight 372.89
Purity >98%
Molecular Weight (g/mol) 372.897
Chemical Name or Material BCTC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.