CAS RN 3644-61-9
CAS RN 3644-61-9
IUPAC Name:
hydrogen 2-methyl-1-(4-methylphenyl)-3-(piperidin-1-yl)propan-1-one chloride
Synonyms:
hydrogen tolperisone chloride
Molecular Weight (g/mol):
281.82
Molecular Formula:
C16H24ClNO
InChi Key:
ZBUVYROEHQQAKL-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CC(CN1CCCCC1)C(=O)C1=CC=C(C)C=C1
1
–
2
of
2
results
1
Tolperisone Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Tolperisone hydrochloride, MedChemExpress
MedChemExpress Tolperisone hydrochloride is a centrally acting muscle relaxant, is indicated for use in the treatment of pathologically increased tone of the cross-striated muscle caused by neurological diseases (damage of the pyramidal tract, multiple sclerosis, myelopathy, encephalomyelitis) and of spastic paralysis and other encephalopathies manifested with muscular dystonia.