missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tolperisone Hydrochloride, Mikromol™
Description
Tolperisone Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Tolperisone Hydrochloride |
| CAS | 3644-61-9 |
| Analyte or Component Names | Tolperisone Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H23 N O . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2-Methyl-1-(4-methylphenyl)-3-(1-piperidinyl)-1-propanone Hydrochloride (1:1), 1-Propanone, 2-methyl-1-(4-methylphenyl)-3-(1-piperidinyl)-, hydrochloride (9CI), Propiophenone, 2,4'-dimethyl-3-piperidino-, hydrochloride (7CI,8CI), 1-Piperidino-2-methyl-3-(4-methylphenyl)propan-3-one hydrochloride, 2,4'-Dimethyl-3-piperidinopropiophenone hydrochloride, Abbsa, Arantoick, Atmosgen, Besnoline, Biocalm, Isocalm, Kineorl, Menopatol, Metosomin, Midocalm, Minacalm, Muscalm, Mydocalm, N 553, Naismeritin, Tolfree, Tolisartine, Tolperisone hydrochloride, Tolpidol |
| InChI Formula | InChI=1S/C16H23NO.ClH/c1-13-6-8-15(9-7-13)16(18)14(2)12-17-10-4-3-5-11-17;/h6-9,14H,3-5,10-12H2,1-2H3;1H |
| SMILES | Cl.CC(CN1CCCCC1)C(=O)c2ccc(C)cc2 |
| IUPAC Name | 2-methyl-1-(4-methylphenyl)-3-piperidin-1-ylpropan-1-one;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?