missing translation for 'onlineSavingsMsg'
Learn More

Tolperisone hydrochloride, MedChemExpress

Product Code. p-200101597
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16434557

Brand: Medchem Express HYB1139

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tolperisone hydrochloride is a centrally acting muscle relaxant, is indicated for use in the treatment of pathologically increased tone of the cross-striated muscle caused by neurological diseases (damage of the pyramidal tract, multiple sclerosis, myelopathy, encephalomyelitis) and of spastic paralysis and other encephalopathies manifested with muscular dystonia.

Tolperisone hydrochloride is a centrally acting muscle relaxant, is indicated for use in the treatment of pathologically increased tone of the cross-striated muscle caused by neurological diseases (damage of the pyramidal tract, multiple sclerosis, myelopathy, encephalomyelitis) and of spastic paralysis and other encephalopathies manifested with muscular dystonia.

Specifications

Chemical Name or Material Tolperisone hydrochloride
CAS 3644-61-9
Color White
Purity Grade Notes Research
Molecular Formula C16H24ClNO
Quantity 50 mg
Solubility Information DMSO : 100 mg/mL (354.84 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 100 mg/mL (354.84 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C)C=C1)C(C)CN2CCCCC2.[H]Cl
Molecular Weight (g/mol) 281.82
Formula Weight 281.82
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.