accessibility menu, dialog, popup

UserName

CAS RN 35920-39-9

N(R)N(R)O(S)(S)NNOHNH2HONHOH3C

CAS RN 35920-39-9

IUPAC Name: (2S,3S,4R)-5-(6-amino-9H-purin-9-yl)-N-ethyl-3,4-dihydroxyoxolane-2-carboxamide
Synonyms: (2S,3S,4R)-5-(6-aminopurin-9-yl)-N-ethyl-3,4-dihydroxyoxolane-2-carboxamide
Molecular Weight (g/mol): 308.3
Molecular Formula: C12H16N6O4
InChi Key: JADDQZYHOWSFJD-QQIVLSGASA-N
SMILES: CCNC(=O)[C@H]1OC([C@H](O)[C@@H]1O)N1C=NC2=C(N)N=CN=C12
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results