missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5’-Ethylcarboxamido Adenosine, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 5'-Ethylcarboxamido Adenosine |
| CAS | 35920-39-9 |
| Molecular Formula | C12 H16 N6 O4 |
| Quantity | 10 mg |
| Synonym | 5-Ethylcarboxamido Adenosine |
| InChI Formula | InChI=1S/C12H16N6O4/c1-2-14-11(21)8-6(19)7(20)12(22-8)18-4-17-5-9(13)15-3-16-10(5)18/h3-4,6-8,12,19-20H,2H2,1H3,(H,14,21)(H2,13,15,16)/t6-,7+,8-,12+/m0/s1 |
| SMILES | CCNC(=O)[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)ncnc23 |
| IUPAC Name | (2S,3S,4R,5R)-5-(6-aminopurin-9-yl)-N-ethyl-3,4-dihydroxyoxolane-2-carboxamide |
| Molecular Weight (g/mol) | 308.29 |
| Formula Weight | 308.1233 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction