missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NECA

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
High affinity adenosine receptor agonist (Ki values are 6.2, 14, and 20 nM for human A3, A1 and A2A receptors respectively; EC50 = 2.4 μM for human A2B). Inhibits platelet aggregation and is centrally active in vivo.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 308.3 |
| Purity | 98% |
| Molecular Weight (g/mol) | 308.30 |
| Chemical Name or Material | NECA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction