CAS RN 349085-38-7
IUPAC Name:
2-(1,3-dimethyl-2,6-dioxo-2,3,6,7-tetrahydro-1H-purin-7-yl)-N-[4-(propan-2-yl)phenyl]acetamide
Synonyms:
2-(1,3-dimethyl-2,6-dioxopurin-7-yl)-N-(4-isopropylphenyl)acetamide
Molecular Weight (g/mol):
355.4
Molecular Formula:
C18H21N5O3
InChi Key:
HEQDZPHDVAOBLN-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=C(NC(=O)CN2C=NC3=C2C(=O)N(C)C(=O)N3C)C=C1
1
–
3
of
3
results
1
HC 030031, TRC
CAS: 349085-38-7 Molecular Formula: C18 H21 N5 O3 Molecular Weight (g/mol): 355.39 Synonym: 1,2,3,6-Tetrahydro-1,3-dimethyl-N-[4-(1-methylethyl)phenyl]-2,6-dioxo-7H-purine-7-acetamide IUPAC Name: 2-(1,3-dimethyl-2,6-dioxopurin-7-yl)-N-(4-propan-2-ylphenyl)acetamide SMILES: CC(C)c1ccc(NC(=O)Cn2cnc3N(C)C(=O)N(C)C(=O)c23)cc1
2
Tocris Bioscience™ HC 030031
CAS: 349085-38-7 Molecular Formula: C18H21N5O3
3
HC-030031, MedChemExpress
MedChemExpress HC-030031 is a potent and selective TRPA1 inhibitor, which antagonizes AITC- and formalin-evoked calcium influx with IC50s of 6.2±0.2 and 5.3±0.2 μM, respectively.