missing translation for 'onlineSavingsMsg'
Learn More

HC-030031, MedChemExpress

Product Code. 16486298
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486298 10 mg 10mg
16472417 50 mg 50mg
16466626 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486298 Supplier Medchem Express Supplier No. HY15064
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress HC-030031 is a potent and selective TRPA1 inhibitor, which antagonizes AITC- and formalin-evoked calcium influx with IC50s of 6.2±0.2 and 5.3±0.2 μM, respectively.

HC-030031 is a potent and selective TRPA1 inhibitor, which antagonizes AITC- and formalin-evoked calcium influx with IC50s of 6.2±0.2 and 5.3±0.2 μM, respectively.

Specifications

Chemical Name or Material HC-030031
CAS 349085-38-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H21N5O3
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (70.35 mM; Need ultrasonic)
SMILES O=C1C2=C(N=CN2CC(NC3=CC=C(C(C)C)C=C3)=O)N(C)C(N1C)=O
Molecular Weight (g/mol) 355.39
Formula Weight 355.39
Percent Purity 95.91%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Mostrar más Mostrar menos

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.